UserName
Five Things to Consider When Buying Balances and Scales
Featured Products
Balances and Scales
Different scales have varying capacities, from a few grams for precision scales to several tons for industrial scales. Ensure the scale's capacity and readability align with your requirements.
For tasks requiring high precision, choose scales with higher sensitivity. For general purposes or industrial use, a balance between accuracy and capacity might be more important.
Regular calibration is essential to ensure the scale's accuracy and reliability, choose between a scale with internal or external calibration.
For industrial settings, look for scales that are robust and resistant to dust, moisture, and other harsh conditions. For laboratory or kitchen use, ensure the scale has features like spill resistance and easy cleaning.
Lastly evaluate the additional features and connectivity options the scale should offer to meet your needs.
Popular Resources
Top Tips for Lab Weighing with OHAUS
External factors such as temperature fluctuations, air currents, electromagnetic interference, and vibrations can all have an impact on the accuracy of an analytical balance.
28392-analytical-balances-1
Cubis II Ultra-High Resolution Balances
The Cubis II MCA66S showed excellent repeatability values with all three different types of vessels. The measurement time is comparable or slightly better in contrast to the tested competitor.
28392-analytical-balances-2
A&D Advanced-Level Analytical/Precision Balances
All the innovative technologies built into the A&D Apollo are designed to provide functional solutions to the real world problems that users face in laboratories or in factories.
28392-analytical-balances-3
Five Things to Consider When Buying Balances and Scales
Different scales have varying capacities, from a few grams for precision scales to several tons for industrial scales. Ensure the scale's capacity and readability align with your requirements.
28392-image2
Top Brands