CAS RN 4170-24-5
IUPAC Nome:
2-chlorobutanoic acid
Sinonimi:
2-chlorobutanoic acid
Peso molecolare (g/mol):
122.55
Formula molecolare:
C4H7ClO2
InChi Key:
RVBUZBPJAGZHSQ-UHFFFAOYNA-N
SMILES:
CCC(Cl)C(O)=O
1
–
1
di
1
risultati
1
2-Chlorobutyric acid, 97%, Thermo Scientific Chemicals
CAS: 4170-24-5 Formula molecolare: C4H7ClO2 Molecular Weight (g/mol): 122.548 Numero MDL: MFCD00021697 InChI Key: RVBUZBPJAGZHSQ-UHFFFAOYSA-N Sinonimo: 2-chlorobutyric acid,butanoic acid, 2-chloro,butyric acid, 2-chloro,ccris 8992,2-chloro-butyric acid,2-chlorbutyric acid,acmc-209huc,acmc-209lcl,2-chloro-n-butyric acid,butanoic acid,2-chloro PubChem CID: 20137 IUPAC Name: 2-chlorobutanoic acid SMILES: CCC(C(=O)O)Cl