CAS RN 730-40-5
CAS RN 730-40-5
IUPAC Nome:
4-[2-(4-nitrophenyl)diazen-1-yl]aniline
Sinonimi:
4-[2-(4-nitrophenyl)diazen-1-yl]aniline
Peso molecolare (g/mol):
242.24
Formula molecolare:
C12H10N4O2
InChi Key:
UNBOSJFEZZJZLR-UHFFFAOYSA-N
SMILES:
NC1=CC=C(C=C1)N=NC1=CC=C(C=C1)[N+]([O-])=O
1
–
4
di
4
risultati
2
Disperse Orange 3 (Technical Grade), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Disperse Orange 3 (Technical Grade), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
Disperse Orange 3 (Technical Grade), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.