CAS RN 81480-40-2
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
1
di
1
risultati
1
Oleic Acid N-Hydroxysuccinimide, TRC
CAS: 81480-40-2 Formula molecolare: C22 H37 N O4 Molecular Weight (g/mol): 379.53 Sinonimo: (9Z)-9-Octadecenoic Acid 2,5-Dioxo-1-pyrrolidinyl Ester,(Z)-1-[(1-Oxo-9-octadecenyl)oxy]-2,5-pyrrolidinedione,1-[[(9Z)-1-Oxo-9-octadecenyl]oxy]-2,5-pyrrolidinedione,N-Hydroxysuccinimide Ester with Oleic Acid,N-Hydroxysuccinimide Oleate,N-Succinimidyl Oleate,Oleic Acid N-Hydroxysuccinimide Ester,Succinimidyl Oleate IUPAC Name: (2,5-dioxopyrrolidin-1-yl) (Z)-octadec-9-enoate SMILES: CCCCCCCC\C=C/CCCCCCCC(=O)ON1C(=O)CCC1=O