UserName
 Additional Selective and Differential Culture Media
Additional Selective and Differential Culture Media
Selective and differential culture media provide the ideal support for the growth of many cell types. They are available in a variety of forms, including both liquid and dry formulations.
MP Biomedicals™ Glycerol, 80% Sterile Solution
Excellent for freezing bacterial and yeast strains.
MP Biomedicals™ Glycerol, 80% Sterile Solution
Sartorius Microsart™ Media
For microbiological QC in the Pharma and Biotech industries
Ready-to-use, prefilled agar media dish with active lid for microbial limits testing according to the U.S. (USP) and E.U. (EP) Pharmacopeia.
Sartorius Microsart™ Media
Corning™ Lymphocyte Separation Medium
Designed for the in vitro isolation of lymphocytes from diluted whole blood
A sterile filtered, iso-osmotic polysucrose and diatrizoate solution with low viscosity.
Corning™ Lymphocyte Separation Medium
Save Now - Exclusive Deals
Complete Your Order - Great Deals
Corning™ Sterile Cell Strainers
Rapidly isolate primary cells to obtain consistently uniform single-cell suspensions from tissues. Corning™ Sterile Cell Strainers are ideal for use in preparation of flow cytometry samples and can be used with stem cells and primary cells.
Corning™ Sterile Cell Strainers
Fisherbrand™ Cell Scrapers
Easily harvest cells from a variety of tissue culture flasks. Fisherbrand™ Cell Scrapers feature a uniquely designed handle and blade to reduce cell damage during the harvesting process.
Fisherbrand™ Cell Scrapers
Syringeless Filters and Syringe Filters
Remove interfering materials and fine particles prior to injection. Constructed predominantly from PVDF or PTFE, syringeless filters and syringe filters are suitable for both aqueous and organic samples at various stages of your workflow, including prefiltration, sample preparation, sterile filtration, laboratory filtration, and gas venting.
Syringeless Filters and Syringe Filters
FAQ
Common selective and differential culture media are:
  • Mannitol Salt Agar (MSA): Selective due to high NaCl for Staphylococcus. Differential since it contains mannitol and phenol red. Mannitol fermentation turns yellow
  • MacConkey Agar: Selective since it contains bile salts and crystal violet, which inhibit the growth of gram-positive bacteria. Differential since it contains lactose and neutral red. Lactose fermentation turns pink/red
  • Eosin Methylene Blue (EMB) Agar: Selective since it contains eosin Y and methylene blue, which inhibit the growth of gram-positive bacteria. Differential because it contains lactose. Lactose fermentation produces dark colonies with a green sheen
  • Hektoen Enteric (HE) Agar: Selective since it contains bile salts and dyes that inhibit the growth of gram-positive bacteria. Differential since it contains lactose, sucrose, salicin, and ferric ammonium citrate. Lactose/sucrose fermentation turns yellow/salmon; hydrogen sulfide producers form black colonies
  • Xylose Lysine Deoxycholate (XLD) Agar: Selective since it contains sodium deoxycholate, which inhibits the growth of gram-positive bacteria. Differential since it contains xylose, lysine, and ferric ammonium citrate. Xylose fermentation turns yellow; Hydrogen sulfide producers form black precipitates
Mannitol Salt Agar (MSA) is considered both selective and differential due to its unique composition and the way it interacts with different types of bacteria.
Selective Properties:
  • High Concentration of Sodium Chloride (NaCl): MSA contains a high concentration of sodium chloride, usually around 7.5-10%. This high salt concentration inhibits the growth of most bacteria but allows the growth of halotolerant bacteria, particularly members of the genus Staphylococcus. This makes the medium selective for these types of bacteria
Differential Properties:
  • Mannitol and Phenol Red: MSA contains the sugar mannitol and the pH indicator phenol red. When bacteria that can ferment mannitol grow on the agar, they produce acidic byproducts. The phenol red in the medium detects this change in pH
    • Mannitol Fermentation: If mannitol fermentation occurs, the phenol red turns yellow due to the acidic environment
    • No Mannitol Fermentation: If mannitol is not fermented, the phenol red remains red or turns pink due to alkaline byproducts
When selecting additional selective and differential culture media, consider the following:
  1. Determine the specific types of microorganisms you aim to isolate and identify. Different media are designed to select for and differentiate between various bacteria, fungi, or other microorganisms.
  2. Evaluate the inhibitory substances present in the medium. These substances are crucial for suppressing the growth of non-target organisms. Ensure that the inhibitors are appropriate for the microorganisms you wish to exclude.
  3. Consider the biochemical or physiological traits that the medium differentiates. This often involves specific metabolic properties, such as carbohydrate fermentation, enzyme activity, or the production of certain pigments.
  4. Assess the nutritional components of the media to ensure they support the growth of the target organisms.
  5. Check the indicator systems used in the medium, such as pH indicators, chromogenic substrates, or redox indicators. These systems should provide clear and easily interpretable results for distinguishing between different types of microorganisms.
Resources and Related Information
Protein Production in Mammalian Cells
Find information on the FreeStyle™ 293 Expression System - the way to turn mammalian cells into a protein production system.
Protein Production in Mammalian Cells
One Cell Biology
Expedite your search for the right cell biology product. Find information on Gibco Media, FBS, supplements, cells, and reagents for cell culture.
One Cell Biology
Media Mastery for Cell Culture
Find information on everything Cell Culture Related.
Media Mastery for Cell Culture
Top Brands