Learn More
Hepatitis C Virus Core Antigen Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Novus Biologicals NB100-93581PCP
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Hepatitis C Virus Core Antigen Polyclonal antibody specifically detects Hepatitis C Virus Core Antigen in Human samples. It is validated for Western Blot
Specifica
| Hepatitis C Virus Core Antigen | |
| Polyclonal | |
| Western Blot | |
| Core protein p19, HCcAg, HCV core antigen, HCV core protein | |
| Hepatitis C Virus (HCV) core protein (HCV-C1) | |
| 0.1 mL | |
| Infections (Virus Bacteria and Parasites), Virology Bacteria and Parasites | |
| 951475 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| PerCP | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |