missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BAFF/BLyS/TNFSF13B Antibody (belimumab), Alexa Fluor™ 350, Novus Biologicals™
Human Monoclonal Antibody
Marca: Novus Biologicals NBP3-27972AF350
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
BAFF/BLyS/TNFSF13B Monoclonal antibody specifically detects BAFF/BLyS/TNFSF13B in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifica
| BAFF/BLyS/TNFSF13B | |
| Monoclonal | |
| Alexa Fluor 350 | |
| ApoL related ligand TALL-1, B lymphocyte stimulator, BAFFB-cell activating factor, B-cell-activating factor, BLyS, BLYSB-lymphocyte stimulator, CD257, CD257 antigen, Dendritic cell-derived TNF-like molecule, DTL, TALL-1delta BAFF, TALL1Delta4 BAFF, THANK, TNF and ApoL-related leukocyte expressed ligand 1, TNF- and APOL-related leukocyte expressed ligand 1, TNF homolog that activates apoptosis, TNFSF20, tumor necrosis factor (ligand) superfamily, member 13b, tumor necrosis factor (ligand) superfamily, member 20, tumor necrosis factor ligand superfamily member 13B, tumor necrosis factor superfamily, member 13b, tumor necrosis factor-like protein ZTNF4, ZTNF4 | |
| TNFSF13B/ BAFF/ CD257 | |
| 0.1 mL | |
| Apoptosis, Cell Cycle and Replication, Cytokine Research, Immunology | |
| 10673 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| belimumab | |
| 50mM Sodium Borate | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto