Learn More
Thermo Scientific™ Amber Screw Cap Micro Tubes
GREENER_CHOICE
Codice prodotto. p-8630000 Sfoglia Tutto MBP Molecular BioProducts Prodotti
Click to view available options
Capacità (sistema metrico):
0.5 mL
0.5 ml
1.5 mL
1.5 ml
2 mL
2 ml
Forma del fondo:
Autoportante
Conico
Non-skirted
Skirted
Tipo di chiusura:
Attached Cap
Not Included
Screw Cap with O-ring (Attached)
Screw Cap with O-ring (Unattached)
Tappo a vite e o-ring, attaccato
Tappo a vite e o-ring, staccato
Imballaggio:
50 provette e tappi attaccati/sacchetto, 10 sacchetti/unità, 10 unità/scatola
50 provette/sacchetto, 10 sacchetti/unità, 10 unità/contenitore; 500 tappi/sacchetto/unità
50 tubes and attached caps/bag
50 tubes/bag, 10 bags/unit, 10 units/case; 500 caps/bag/unit
500 x pk, 10 pk x cs, 5000 x cs. Single Bagged. ZipLock bag.
Dimensione della confezione:
pezzi 500
pezzi 5000
Codice prodotto. 16245530
Marca: Thermo Scientific™ 3422ANK
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Pure Protection: Prepare, transfer and store samples securely with Thermo Scientific Screw Cap Microtubes. Available in multiple volumes, packaging options and clean claims.
Thermo Scientific™ Screw Cap Micro Tubes robust design and material selection are well suited for work with biologics and offer rich features, including ultra-clear externally threaded tubes and UV protective amber tubes. Options are available with or without a self-standing skirted bottom, non-knurl and knurled tubes for single hand operation, in 0.5mL, 1.5mL and 2.0mL sizes, and the option of multicolored caps, with or without tethered straps. Screw Cap Micro Tube Caps offer a durable design incorporating an O-ring that helps prevent leaks under demanding conditions.
Note: For screw caps, see Cat. No. 3471AS.
TRUSTED_SUSTAINABILITY
Specifica
Autoclavabile Not Autoclavable
Capacità (sistema metrico) 0.5 mL
Colore Amber
Materiale Polypropylene (Virgin)
Include NonKnurl
Imballaggio 500 x pk, 10 pk x cs, 5000 x cs. Single Bagged. ZipLock bag.
Temperature Range (Metric) Cold Storage: -80° to +95°C. Liquid Nitrogen: Short term Vapor Phase & Quick Flash Freeze acceptable, not recommmended for LN2 submersion storage
Dichiarazioni di pulizia Non-Sterile Free of: Rnase/Dnase, Human and Mouse DNA, Pyrogen & Endotoxin, ATP, PCR Inhibitor
Resistenza UV (UV) Ultra violet protection avg. 1.22% transmittance between 220 and 500nm
Proprietà di legame Low Nucleic Acid and Protein Binding & Low Enzyme Inhibition
Coppia max Manual/Hand Torque: Standard: 0.125 lbsf.in to 2.5 lbsf.in; Metric: 1.412 Ncm to 28.24 Ncm. Automated/Robotic Torque Standard: 1.312 lbsf.in to 2.5 lbsf.in; Metric: 14.83 Ncm to 28.24 Ncm
Sterilità Non-sterile
Graduated No
Tipo di chiusura Not Included
Fit for Purpose PCR Ready
Forma del fondo Skirted
RCF max 18,000 x g
Product Type Screw-cap Microtube
† Subject to Thermo Fisher Scientific standard limited warranty. See thermoscientific.com or your sales representative for details.
Screw cap micro tubes and caps are not recommended for autoclave. Sterile options available.
Not intended to submerge and store in liquid phase LN2.