Learn More
1,1-Di(tert-butylperoxy)cyclohexane, 50% solution in mineral oil
Codice prodotto. 10582802 Sfoglia Tutto Thermo Scientific Chemicals Prodotti
Click to view available options
Quantità:
10 mL
250 mL
Dimensione della confezione:
10mL
250mL
Codice prodotto. 10582802
Marca: Thermo Scientific Acros 361312500
Richiesta di formato massivo o personalizzato
CAS: 3006-86-8 | C14H28O4 | 260.374 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Radical initiators
TRUSTED_SUSTAINABILITY
Specifica
Materiale o nome chimico 1, 1-Di(tert-butylperoxy)cyclohexane
Colore Undesignated
Nota nome 50% Solution in Mineral Oil
CAS 3006-86-8
CAS min. % 49.0
CAS max % 51.0
Formula molecolare C14H28O4
Numero MDL MFCD00084800
Beilstein 07, III, 30
Densità 0.9500g/mL
Sinonimo 1,1-bis tert-butylperoxy cyclohexane, 1,1-di tert-butylperoxy cyclohexane, unii-jk62j3e3q4, 1,1-bis t-butylperoxy cyclohexane, peroxide, 1,1'-cyclohexylidenebis 2-1,1-dimethylethyl, perhexa-c, cyclohexylidenebis tert-butyl peroxide, peroxide, cyclohexylidenebis 1,1-dimethylethyl, 1,1-di t-butylperoxy-cyclohexane
InChI Key HSLFISVKRDQEBY-UHFFFAOYSA-N
SMILES CC(C)(C)OOC1(CCCCC1)OOC(C)(C)C
IUPAC Name 1,1-bis(tert-butylperoxy)cyclohexane
Molecular Weight (g/mol) 260.374
PubChem CID 76359
Peso formulazione 260.37
Percent Purity 49 to 51%
Imballaggio Plastic bottle
Quantità 250 mL
Forma fisica Liquid