Learn More
Trajan Scientific and Medical™ 1/32 in. O.D. PEEKsil™ Tubing
Click to view available options
Overall Length (Metric):
50 mm
100 mm
150 mm
250 mm
500 mm
Diametro (sistema metrico) interno:
0.025 mm
0.05 mm
0.075 mm
0.1 mm
0.15 mm
Dimensione della confezione:
Pezzo
pezzi 2
Descrizione
- Fused silica internal wall provides smooth surface, free of imperfections
- PEEKsil compatible with most organic solvents and resistant to strong acids
- Effective pH range of 0 to 10
- Flexibility ensures easy connection between columns, detectors and injectors
Specifica
Specifica
| Tipo | Tubing |
| Include | 2/Pk. |
| Diametro (sistema metrico) interno | 0.1 mm |
| Overall Length (Metric) | 50 mm |