Learn More
Trajan Scientific and Medical™ 1/32 in. O.D. PEEKsil™ Tubing
Codice prodotto. p-7114374
Click to view available options
Overall Length (Metric):
50 mm
100 mm
150 mm
250 mm
500 mm
Diametro (sistema metrico) interno:
0.025 mm
0.05 mm
0.075 mm
0.1 mm
0.15 mm
Dimensione della confezione:
Pezzo
pezzi 2
Codice prodotto. 15208753
Marca: Trajan Scientific and Medical™ 0624245
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
A mechanically robust polymer-sheathed fused silica tubing for use in HPLC systems
  • Fused silica internal wall provides smooth surface, free of imperfections
  • PEEKsil compatible with most organic solvents and resistant to strong acids
  • Effective pH range of 0 to 10
  • Flexibility ensures easy connection between columns, detectors and injectors
    Specifica
    Tipo Tubing
    Include 2/Pk.
    Diametro (sistema metrico) interno 0.025 mm
    Overall Length (Metric) 250 mm