Learn More
Sartorius™ Standard Flexboy™ Bioprocessing Bags
Provides a single-use alternative to glass and plastic carboys in bioprocess applications. Sartorius™ Standard Flexboy™ Bioprocessing Bags feature EVA tubing and MPC connections, and are extensively qualified to be safe and robust. Pre-sterilized, ready-to-use bags with volumes between 5 and 50L.
Marca: Sartorius™ FFB102340
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Superior biopharmaceutical fluid handling- Ideal for the preparation, storage and transport of biopharmaceuticals, intermediates and final bulk products
- The broad chemical compatibility assures the safe processing of a wide range of process fluids
- Single-use alternative to traditional glass, stainless steel and rigid plastic carboys for many applications
- Ethyl Vinyl Acetate (EVA) tubing for superior flexibility, impact strength and low-temp qualities
- Three bag ports per bag with easy-to-use and secure MPC male couplings:
- Bag ports 1 and 2: 3/8 in. x 15/32 in. x 10cm with dust caps and slide clamps
- Bag port 3: 3/16 in. x ¼ in. x 5cm with a septum
- 100% integrity testing of bag and connections to ensure process safety and integrity
- Full compliance with ISO 11137 for the highest sterility assurance level
- Single-use system reduces the risk of cross contamination between batches and products
- Costly and time-consuming CIP and SIP operations are minimized