Learn More
Integra Biosciences™ 1-Channel Adapter
Click to view available options
Tipo:
Adapter for Disposable Tips
Adapter with Ejector for GripTips
Adapter, 150 mm
Adapter, 40 mm
Dimensione della confezione:
Pezzo
pezzi 5
Descrizione
One-channel adaptor for use with VACUBOY hand operator.
The VACUBOY hand operator can be used with VACUSAFE comfort vacuum system. Enables liquid residues to be disposed of reliably, precisely and conveniently:
The VACUBOY hand operator can be used with VACUSAFE comfort vacuum system. Enables liquid residues to be disposed of reliably, precisely and conveniently:
- With a precisely dosed volume
- Without loss of drops
- Without risk of contamination
- Without causing tiredness
- Energy saving, the manual operator - the heart of the system - can be operated as a single or 8 channel aspirator
- All parts in contact with fluids are autoclavable
Specifica
Specifica
| Descrizione | 1-channel stainless steel |
| Tipo | Adapter, 150 mm |
| Da utilizzare con (apparecchiatura) | VACUBOY Hand Operator |