Learn More
1-Chloro-3-iodobenzene, 97%, Thermo Scientific™
Product Code. 11310857
Change view
Click to view available options
Quantity:
25 g
50 g
100 g
Unit Size:
100g
25g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11310857 50 g 50g
10673245 25 g 25g
10351615 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11310857 Supplier Thermo Scientific Maybridge Supplier No. TL00516EE
 Restricted Item
Spedizione stimata: 30-11-2026
Add to basket
Chemical Identifiers
CAS 625-99-0
Molecular Formula C6H4ClI
Molecular Weight (g/mol) 238.45
MDL Number MFCD00001046
InChI Key JMLWXCJXOYDXRN-UHFFFAOYSA-N
Synonym 3-chloroiodobenzene, m-chloroiodobenzene, benzene, 1-chloro-3-iodo, 3-iodochlorobenzene, 3-chloro-1-iodobenzene, 1-chloro-3-iodo-benzene, m-chlorophenyl iodide, m-iodochlorobenzene, metachloroiodobenzene, 3-chloro-iodobenzene
PubChem CID 12269
IUPAC Name 1-chloro-3-iodobenzene
SMILES ClC1=CC=CC(I)=C1
Specifications
Density 1.9200g/mL
Assay Percent Range 97%
Packaging Amber glass bottle
Linear Formula ClC6H4I
Quantity 50 g
Formula Weight 238.45
Percent Purity 97%
Chemical Name or Material 1-Chloro-3-iodobenzene