Learn More
Wheaton™ 1 μL Inoculation Loop
Inoculation loop calibrated to deliver a known volume of fluid sample to enable accurate microbiological analysis.
Marca: Wheaton™ 711310
346.21 EUR valido fino al 2024-12-31
Usa il codice promo "23259" per avere il tuo prezzo promozionale.
Dettagli aggiuntivi : Peso : 9.89500kg
Descrizione
- Meets quantitative standards for management of microbiological samples
- For accurate liquid transfer and plating
- Calibrated
Specifica
Yes | |
ABS | |
1 μL | |
Blue | |
Packaged in peel pouch | |
No |
Sterile | |
199 mm | |
Standard | |
6000 | |
Yes |