Learn More
Masterflex™ Rubinetto d'arresto a 1 via
Regulate flow without any leaking using these fittings. Cole-Parmer™ Stopcocks with Luer Connections are available with either a male luer lock or a male luer slip.
Marca: Masterflex™ 30600-00
Dettagli aggiuntivi : Peso : 0.02520kg
Descrizione
Regulate flow without any leaking using these fittings. Cole-Parmer™ Stopcocks with Luer Connections are available with either a male luer lock or a male luer slip.
- Handle rotates 90°
- PC body and HDPE stem
Specifica
Valvola di chiusura | |
Policarbonato e HDPE | |
Luer |
Policarbonato e HDPE, HDPE | |
Male Luer Lock or A Male Luer Slip |