Learn More
Thermo Scientific™ Ghiere in grafite 100% per strumenti Thermo Scientific
Codice prodotto. 10409624 Sfoglia Tutto Thermo Scientific Prodotti
Click to view available options
Da utilizzare con (apparecchiatura):
Agilent Injectors and Non-MS Detectors
Strumenti Thermo Scientific
Thermo Scientific TRACE Ultra SSL Injectors and Non-MS Detectors
Diametro (sistema metrico):
0.32 mm
0.53 mm
3.175 mm
6.35 mm
Da 0.1 a 0.25 mm
Dimensione della confezione:
pezzi 10
Codice prodotto. 10409624
Marca: Thermo Scientific™ 290GA107
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Ensure reliable connections for Thermo Scientific™ GC instruments with these Thermo Scientific™ 100% Graphite Ferrules for Thermo Scientific Instruments.
Ensure optimal performance of your Thermo Scientific™ GC instrument with Thermo Scientific™ Ferrules. They are available in three different materials and in various dimensions to accommodate a range of instruments and applications. All ferrules are are supplied in contaminant-free, individual blister packs.
Features
  • Available in 100% graphite, 15% graphite/85% vespel and SilTite metal
  • Ferrules for different types of injectors and detectors
TRUSTED_SUSTAINABILITY
Specifica
Tipo Packed Column
Descrizione 1/4 in. O.D.; Agilent 5890, 6890, 6850
Diametro (sistema imperiale) 1/4 in.
Diametro (sistema metrico) 6.35 mm
Da utilizzare con (apparecchiatura) Agilent Injectors and Non-MS Detectors
Materiale 100% Graphite