Learn More
Stainless Steel, Nitronic 50, S20910, 100g, ARMI∣MBH
Codice prodotto. 18031190
Click to view available options
Quantità:
100 g
Dimensione della confezione:
100g
Codice prodotto. 18031190
Marca: ARMI/MBH IARMFEN5018C
ARMI∣MBH produces high-quality metal alloy certified reference materials and set up standards for Spark-OES, ICP, and XRF elemental analysis. The offering includes steel, aluminum, titanium, nickel, cobalt, copper, magnesium, and more.
Stainless Steel, Nitronic 50, S20910, 100g
TRUSTED_SUSTAINABILITY
Specifica
Materiale o nome chimico Aluminium/Oxygen/Tin/Phosphorus/Niobium/Cobalt/Nitrogen/Vanadium/Molybdenum/Carbon/Copper/Silicon/Manganese/Nickel/Tungsten/Chromium/Bismuth/Boron/Cadmium/Calcium/Lead/Magnesium/Selenium/Tantalum/Zinc/Zirconium/Antimony/Arsenic/Titanium
CAS 7439-89-6
Quantità 100 g
Forma Pieces
Concentration or Composition (by Analyte or Components) Aluminium 0.004 %/Oxygen 0.004 %/Tin 0.005 %/Phosphorus 0.016 %/Niobium 0.019 %/Cobalt 0.029 %/Nitrogen 0.031 %/Vanadium 0.046 %/Molybdenum 0.104 %/Carbon 0.168 %/Copper 0.222 %/Silicon 0.32 %/Manganese 0.429 %/Nickel 2 %/Tungsten 2.63 %/Chromium 12.4 %/Bismuth (<0.005) %/Boron (<0.005) %/Cadmium (<0.005) %/Calcium (<0.005) %/Lead (<0.005) %/Magnesium (<0.005) %/Selenium (<0.005) %/Tantalum (<0.005) %/Zinc (<0.005) %/Zirconium (<0.005) %/Antimony (<0.05) %/Arsenic (0.003) %/Titanium (0.004) %