missing translation for 'onlineSavingsMsg'
Learn More

Tool Steel, AISI L6, T61206, 100g, ARMI∣MBH

Codice prodotto. 17933026
Click to view available options
Quantità:
100 g
Dimensione della confezione:
100g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Codice prodotto. 17933026

Marca: ARMI/MBH IARMFEL618C

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

Les retours ne sont pas autorisés pour ce produit. View return policy

ARMI∣MBH produces high-quality metal alloy certified reference materials and set up standards for Spark-OES, ICP, and XRF elemental analysis. The offering includes steel, aluminum, titanium, nickel, cobalt, copper, magnesium, and more.

Tool Steel, AISI L6, T61206, 100g
TRUSTED_SUSTAINABILITY

Specifications

Materiale o nome chimico Tungsten/Tin/Zinc/Magnesium/Boron/Oxygen/Sulfur/Niobium/Arsenic/Titanium/Nitrogen/Cobalt/Vanadium/Phosphorus/Aluminium/Copper/Molybdenum/Silicon/Manganese/Chromium/Carbon/Nickel/Iron
CAS 7439-89-6
Quantità 100 g
Forma Pieces
Concentration or Composition (by Analyte or Components) Tungsten -30 ppm/Tin -20 ppm/Zinc -20 ppm/Magnesium -7 ppm/Boron 0.0008 %/Oxygen 0.0012 %/Sulfur 0.0017 %/Niobium 0.003 %/Arsenic 0.0033 %/Titanium 0.0033 %/Nitrogen 0.0064 %/Cobalt 0.007 %/Vanadium 0.008 %/Phosphorus 0.013 %/Aluminium 0.02 %/Copper 0.027 %/Molybdenum 0.237 %/Silicon 0.264 %/Manganese 0.605 %/Chromium 0.71 %/Carbon 0.718 %/Nickel 1.34 %/Iron (96.3) %
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.