missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brexpiprazole, MedChemExpress
Click to view available options
Quantità:
5 mg
10 mg
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
10mg
200mg
500mg
50mg
5mg
Description
Brexpiprazole (OPC-34712), an atypical orally active antipsychotic drug, is a partial agonist of human 5-HT1A and dopamine D2L receptor with Kis of 0.12 nM and 0.3 nM, respectively. Brexpiprazole is also a 5-HT2A receptor antagonist with a Ki of 0.47 nM. Brexpiprazole also shows potent antagonist activity at human noradrenergic α1B (Ki=0.17 nM) and α2C receptors (Ki=0.59 nM).
Specifications
Specifications
| Materiale o nome chimico | Brexpiprazole |
| CAS | 913611-97-9 |
| Colore | White |
| Note sul grado di purezza | Research |
| Formula molecolare | C25H27N3O2S |
| Quantità | 100 mg |
| Sinonimo | OPC-34712 |
| Informazioni di solubilità | DMSO : 25 mg/mL (57.66 mM; ultrasonic and warming and heat to 80°C) |
| SMILES | O=C1NC2=C(C=CC(OCCCCN3CCN(C4=C(C=CS5)C5=CC=C4)CC3)=C2)C=C1 |
| Molecular Weight (g/mol) | 433.57 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction