missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IFN-beta Antibody, R&D Systems™

Mouse Monoclonal Antibody
Marca: R&D Systems MAB8141-100
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
IFN-beta Monoclonal specifically detects IFN-beta in Human samples. It is validated for Neutralization, Immunocytochemistry.
Specifica
| IFN-beta | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human IFN-beta in direct ELISAs. | |
| Human | |
| Supernatant |
| Neutralization, Immunocytochemistry | |
| 937912 | |
| Neutralization 0.2-1.2 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| Fibroblast interferon, IFB, IFBIFNB, IFF, IFNB, IFNB1, IFNbeta, IFN-beta, interferon beta, interferon, beta 1, fibroblast, MGC96956 | |
| Chinese hamster ovary cell line CHO-derived recombinant human IFN-beta | |
| 100 μg | |
| 3456 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto