Learn More
Carbonic Anhydrase IX/CA9 Antibody (09) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Marca:  Novus Biologicals NBP3-26103
Codice prodotto. 30038881
  • 500.00€ / 100µL
Questo articolo non è restituibile. Consulta la politica di reso
Descrizione
Descrizione
Carbonic Anhydrase IX/CA9 Monoclonal antibody specifically detects Carbonic Anhydrase IX/CA9 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifica
Specifica
Carbonic Anhydrase IX/CA9
Monoclonal
Unconjugated
0.2 um filtered solution in PBS
Mouse
Protein A purified
RUO
Primary
Human
Purified
Immunohistochemistry, Immunohistochemistry (Paraffin)
09
Immunohistochemistry, Immunohistochemistry-Paraffin 1:100-1:500
CAIX, CA-IX, Carbonate dehydratase IX, carbonic anhydrase 9, carbonic anhydrase IX, carbonic dehydratase, EC 4.2.1.1, G250, Membrane antigen MN, MN, P54/58N, PMW1, RCC-associated antigen G250, RCC-associated protein G250, Renal cell carcinoma-associated antigen G250
Hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Carbonic Anhydrase IX/CA9 (Accession#: NP_001207.2, Met1-Asp414)
100 μL
Cancer, HIF Target Genes, Hypoxia, Lipid and Metabolism, Metastasis, Renal Cell Carcinoma Cell Markers
768
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG1