Learn More
Hormone-sensitive Lipase/HSL, p Ser660 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Novus Biologicals NBP3-05459
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Hormone-sensitive Lipase/HSL Polyclonal antibody specifically detects Hormone-sensitive Lipase/HSL in Human, Mouse samples. It is validated for Western Blot
Specifica
| Hormone-sensitive Lipase/HSL | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000 | |
| hormone-sensitive lipase, hormone-sensitive lipase testicular isoform, HSLEC 3.1.1.79, LHS, lipase, hormone-sensitive | |
| A synthetic phosphorylated peptide around S660 of human Hormone-sensitive Lipase/HSL (NP_005348.2). QTSRS | |
| 100 μg | |
| Cancer, Cholesterol Metabolism, Diabetes Research, Lipid and Metabolism, Lipid Droplets, Phospho Specific, Signal Transduction | |
| 3991 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |