Learn More
MP Biomedicals™ 12mm Culture Tubes
Sfoglia Tutto MP Biomedicals Prodotti
Click to view available options
:
Case
Descrizione
- 12mm x 75mm, borosilicate glass, disposable multipurpose culture tubes.
- Packaged in shrink-wrapped trays of 250 to prevent contamination or breakage.
- Ideal for tissue culture and clinical chemistry
Specifica
Specifica
| Descrizione | 12mm x 75mm, borosilicateglass, disposable multipurpose culture tubes |
| Quantità | Case |
| Diametro (sistema metrico) | 12 mm |