Learn More
Keystone Cleanroom Products 1421B Tyvek™ Autoclave Sterilization Wrapper
GREENER_CHOICE
Codice prodotto. 30020285 Sfoglia Tutto Keystone Cleanroom Products Prodotti
Click to view available options
Dimensioni (L x P):
1.5 x 1.5 in.
10 x 10 in.
11 x 9 in.
12 x 12 in.
15 x 15 in.
18 x 18 in.
2 x 2 in.
20 x 20 in.
22 x 18 in.
24 x 24 in.
3 x 3 in.
30 x 15 in.
30 x 30 in.
36 x 36 in.
4 x 4 in.
40 x 40 in.
45 x 45 in.
48 x 48 in.
5 x 5 in.
54 x 54 in.
6 x 6 in.
7.5 x 7.5 in.
72 x 54 in.
8 x 8 in.
80 x 40 in.
9 x 9 in.
Dimensione della confezione:
pezzi 1000
pezzi 125
pezzi 2000
pezzi 250
pezzi 50
pezzi 500
Codice prodotto. 30020285
Marca: Keystone Cleanroom Products 70TK5454
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Autoclavable Wrapper provides an excellent bacterial and microbial barrier for superior performance post steam sterilization.
Tyvek™ Autoclavable Wrappers are breathable while providing an excellent bacterial and microbial barrier for superior performance post steam sterilization.
  • Lint-free (Helmke Drum) means no airborne particulates are generated during its use as sterilization packaging
  • Clean and consistently cut using an automated CAD system per ISO 8 Requirements
  • Packaged in an ISO 6 Controlled Clean Room
  • Designed for Steam Sterilization
  • Hydrophobic
  • Resists tearing and punctures
Specifica
Autoclavabile Autoclavable
Certificazione/Conformità Certificate of Conformance (COC)
Tipo di chiusura None
Colore White
Monouso Single-use
Da utilizzare con (applicazione) Steam Sterilization
Da utilizzare con (apparecchiatura) Pharmaceutical Equipment and Components
Materiale Tyvek
Sterilità Non-sterile
Intervallo di temperatura (sistema imperiale) Up to 250°F
Temperature Range (Metric) Up to 125°C
Dimensioni (L x P) 54 x 54 in.
Lunghezza (sistema imperiale) 54 in.
Lunghezza (sistema metrico) 137 cm
Larghezza (sistema imperiale) 54 in.
Larghezza (sistema metrico) 137 cm
Product Type Sterilization Wrap