Learn More
Keystone Cleanroom Products 1422A Tyvek™ Self Seal Autoclave Bag with Integrated Mittens, See Through Window, and Steam Indicator
Sfoglia Tutto Keystone Cleanroom Products Prodotti
Click to view available options
Dimensioni (L x P):
22 x 20 in.
Dimensione della confezione:
pezzi 100
Descrizione
- Tyvek™/PE Film construction for excellent strength and exceptional microbial barrier
- Designed for steam sterilization
- Easy to use self-sealing closure system
- Integrated Mittens
- Recommended for sterilization of pharmaceutical equipment
- Manufactured and packaged in controlled cleanroom environment
- Make aseptic connections inside the Isolator Bag
- Eliminates risks of contamination
Specifica
Specifica
| Autoclavabile | Autoclavable |
| Capacità (sistema imperiale) | 1217 fl. oz. |
| Certificazione/Conformità | COC |
| Tipo di chiusura | Self-adhesive |
| Colore | White |
| Monouso | Single-use |
| Da utilizzare con (applicazione) | Steam Sterilization |
| Materiale | Tyvek |
| Sterilità | Non-sterile |
| Senza lattice | Yes |
| Vedi altri risultati |