Learn More
Corning™ 1536-Well White Polystyrene Not Treated Microplate, No Lid, With Generic Bar Code, Non-sterile
Sfoglia Tutto Corning Prodotti
Click to view available options
No. of Wells:
1536
Dimensione della confezione:
pezzi 50
Descrizione
- Labels applied to all 4 sides of the plate
- No sequence number duplicates
- Exceptional print quality and resolution
- Resistant to commonly used laboratory reagents
- Resistant to temperature ranges of -80ºC to +52ºC
Microplate Features:
- Excellent signal dynamic range due to low background fluorescence and enhanced signal intensity
- Superior flatness -minimizes variability within a plate and between plates
- Flat bottom wells are compatible with automated dispensers including the PerkinElmer FlexDrop™
- Not treated (or medium binding) polystyrene surface is hydrophobic in nature and binds biomolecules through passive interactions.
- Suitable primarily for the immobilization of large molecules, such as antibodies, that have large hydrophobic regions that can interact with the surface (binding capacity of approximately 100 to 200 ng IgG/cm²)
Specifica
Specifica
| Codice a barre | Includes Barcode |
| Coperchio | Without Lid |
| No. of Wells | 1536 |
| Volume (sistema metrico) di lavorazione | Up to 8 μL |
| Sterilità | Non-sterile |
| Trattamento superficiale | Non trattata |
| Quantità | 10/Pk., 50/Cs. |