Learn More
1H,1H,2H,2H-Perfluorohexanesulfonic acid 100 μg/mL in Methanol:Water, Dr. Ehrenstorfer
Codice prodotto. 30310138
Click to view available options
Quantità:
1 mL
Dimensione della confezione:
1mL
Codice prodotto. 30310138
Marca: Dr. Ehrenstorfer DREA15986903MW100
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifica
Quantità 1 mL
Concentrazione 100 μg/mL
Certificazione/Conformità ISO 17034
Solvente o matrice Methanol/Water