missing translation for 'onlineSavingsMsg'
Learn More

Dichloromethane, Analytical Grade, Penta Chemicals

Codice prodotto. 30020676
Click to view available options
Quantità:
1 L
2.5 L
Dimensione della confezione:
1L
2.50L
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Codice prodotto. 30020676

Marca: PENTA CHEMICALS 1238011000

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Top quality chemicals for laboratories, schools, hospitals, research institutes, and large manufacturing plants.

About us

The company PENTA was established in 1990 under the trade name Ing. Petr Švec - PENTA s.r.o.. From the very beginning, we have been a solely Czech company which is managed by the Švec family, including company founder Petr Švec.

We specialize in the manufacture and distribution of pure, laboratory, pharmaceutical and specialty chemicals, which we market under the trade name Penta. We manufacture and supply a wide range of organic and inorganic chemicals, in large volumes, with a quality ranging from technical, to pure, to A.G. and ACS.

High quality and flexibility

Thanks to the high quality of our products, and our ability to fulfil specific customer requirements, we rank among the leading suppliers of qualified chemicals in the domestic market, and more recently we have also expanded into the global market.

We manufacture the chemicals in our own modern complex, which fulfils the strictest quality and safety criteria. Our large warehouse facilities, including two automated warehouses, enable us to accomodate client requirements. We package the products in accordance with all legislative requirements, and can deliver them to the customer using our own ADR transport on request.

Environmental responsibility

Our priority is to supply the maximum quality of chemicals, reliability, and environmental awareness. Our efforts are evidenced by many awards and certificates, including ISO 9001, ISO 14001 and the Responsible Care certificate for responsible business in the chemical industry.

Identificatori chimici

Molecular Weight (g/mol) 84.93

Specifica

Quantità 1 L
Percent Purity ≥99.5%
Grado Analytical
compliance-icons compliance-icons
Identificativo prodotto
  • Dichloromethane
Avvertenza
  • Attenzione
Categoria di pericolo
  • cancerogenicità Categoria 2
  • Lesione oculare grave/irritazione oculare Categoria 2
  • Corrosione/irritazione della pelle Categoria 2
  • Tossicità specifica per organi bersaglio (esposizione ripetuta) Categoria 2
  • Tossicità specifica per organi bersaglio (esposizione singola) Categoria 3
Indicazioni di pericolo
  • H315-Provoca irritazione cutanea.
  • H319-Provoca grave irritazione oculare.
  • H335-Può irritare le vie respiratorie.
  • H336-Può provocare sonnolenza o vertigini.
  • H351-Sospettato di provocare il cancro.
  • H373-Può provocare danni agli organi.
Consigli di prudenza
  • P201-Procurarsi istruzioni specifiche prima dell'uso.
  • P261-Evitare di respirare la polvere/i fumi/i gas/la nebbia/i vapori/gli aerosol.
  • P280-Indossare guanti/indumenti protettivi/Proteggere gli occhi/il viso.
  • P305+P351+P338-IN CASO DI CONTATTO CON GLI OCCHI: sciacquare accuratamente per parecchi minuti. Togliere le eventuali lenti a contatto se è agevole farlo. Continuare a sciacquare.
  • P308+P313-IN CASO di esposizione o di possibile esposizione, consultare un medico.
Informazioni supplementari
  • MIXTURE LIST-Contiene: Dichloromethane
Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato