Learn More
2,3,4-Tri-O-benzyl-beta-L-arabinopyranose, 97%, Thermo Scientific™
Product Code. 11309755
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11309755 250 mg 250mg
11399745 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11309755 Supplier Thermo Scientific Alfa Aesar Supplier No. L19790.MD
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
2,3,4-Tri-O-benzyl-beta-L-arabinopyranose is used as a pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 77943-33-0
Molecular Formula C26H28O5
Molecular Weight (g/mol) 420.505
MDL Number MFCD05664044
InChI Key HTSKDJMXBBFKKG-SSUZURRFSA-N
Synonym 2,3,4-tri-o-benzyl-beta-l-arabinopyranose, 2,3,4-tri-o-benzyl-b-l-arabinopyranose, 2s,3r,4s,5s-3,4,5-tris benzyloxy oxan-2-ol, 2s,3r,4s,5s-3,4,5-tris benzyloxy tetrahydro-2h-pyran-2-ol
PubChem CID 74892353
IUPAC Name (2S,3R,4S,5S)-3,4,5-tris(phenylmethoxy)oxan-2-ol
SMILES C1C(C(C(C(O1)O)OCC2=CC=CC=C2)OCC3=CC=CC=C3)OCC4=CC=CC=C4
Specifications
Melting Point 76°C to 88°C
Quantity 250 mg
Solubility Information Insoluble in water.
Optical Rotation +78° (c=1 in Methanol)
Formula Weight 420.51
Percent Purity 97%
Chemical Name or Material 2,3,4-Tri-O-benzyl-beta-L-arabinopyranose