Learn More
2-Bromo-5-(trifluoromethyl)aniline, 97%
Product Code. 11413408
Change view
Click to view available options
Quantity:
1 g
25 g
Unit Size:
1g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11413408 25 g 25g
11493398 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11413408 Supplier Thermo Scientific Alfa Aesar Supplier No. A17810.14
 Restricted Item
Spedizione stimata: 02-12-2026
Add to basket
CAS: 454-79-5 | C7H5BrF3N | 240.023 g/mol
2-Bromo-5-(trifluoromethyl)aniline is used in the synthesis and biochemical evaluation of series of inhibitors of hepatitis C virus (HCV) NS3 protease.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Bromo-5-(trifluoromethyl)aniline is used in the synthesis and biochemical evaluation of series of inhibitors of hepatitis C virus (HCV) NS3 protease.
Solubility
Not miscible or difficult to mix in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed in a cool, dry and well ventilated place.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 454-79-5
Molecular Formula C7H5BrF3N
Molecular Weight (g/mol) 240.023
MDL Number MFCD00007637
InChI Key PZDVFXUBTKPFSG-UHFFFAOYSA-N
Synonym 2-bromo-5-trifluoromethyl aniline, 3-amino-4-bromobenzotrifluoride, 2-bromo-5-trifluoromethyl benzenamine, 2-bromo-5-trifluoromethylphenylamine, 2-amino-1-bromo-4-trifluoromethylbenzene, 2-amino-4-trifluoromethylbromobenzene, 6-bromo-alpha,alpha,alpha-trifluoro-m-toluidine, 3-amino-4-bromo trifluoromethyl benzene, benzenamine, 2-bromo-5-trifluoromethyl, 4-bromo-3-aminobenzotrifluoride
PubChem CID 67989
IUPAC Name 2-bromo-5-(trifluoromethyl)aniline
SMILES C1=CC(=C(C=C1C(F)(F)F)N)Br
Specifications
Density 1.675
Boiling Point 81°C to 82°C (5 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.522
Quantity 25 g
UN Number UN2810
Beilstein 2094535
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 240.03
Percent Purity 97%
Chemical Name or Material 2-Bromo-5-(trifluoromethyl)aniline