Learn More
2-Methyl-1-phenyl-2-propanol, TRC
Specifica
Specifica
| CAS | 100-86-7 |
| Molecular Formula | C10H14O |
| Quantity | 5 g |
| Synonym | alpha,alpha-Dimethylbenzeneethanol, 1,1-Dimethyl-2-phenylethanol, 1,1-Dimethyl-2-phenylethyl Alcohol, 1,1-Dimethylphenylethanol;1-Benzyl-1-methylethanol, 1-Phenyl-2-hydroxy-2-methylpropane, 1-Phenyl-2-methylpropan-2-ol, 2-Benzyl-2-propanol;2-Hydroxy-2-methyl-1-phenyl-1-propane, 2-Hydroxy-2-methyl-1-phenylpropane, 2-Methyl-1-phenyl-2-propanol, 2-Methyl-3-phenyl-2-propanol, Benzyldimethylcarbinol, DMBC, Dimethylbenzylcarbinol, NSC 27228, NSC 46103, NSC 5236, Phenyl-tert-butanol, alpha,alpha-Dimethylphenethanol, alpha,alpha-Dimethylphenethyl Alcohol; |
| InChI Formula | InChI=1S/C10H14O/c1-10(2,11)8-9-6-4-3-5-7-9/h3-7,11H,8H2,1-2H3 |
| SMILES | CC(C)(O)Cc1ccccc1 |
| IUPAC Name | 2-methyl-1-phenylpropan-2-ol |
| Molecular Weight (g/mol) | 150.22 |
| Formula Weight | 150.1045 |
| Chemical Name or Material | 2-Methyl-1-phenyl-2-propanol |