missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-5-(trifluoromethoxy)benzamide, 97%, Thermo Scientific™

Codice prodotto. 15446357
Click to view available options
Quantità:
1 g
5 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 1261828-46-9
Formula molecolare C9H8F3NO2
Molecular Weight (g/mol) 219.163
Numero MDL MFCD18399714
InChI Key JHVXNWZOAYHTHU-UHFFFAOYSA-N
Sinonimo 2-methyl-5-trifluoromethoxy benzamide
PubChem CID 68318548
IUPAC Name 2-methyl-5-(trifluoromethoxy)benzamide
SMILES CC1=C(C=C(C=C1)OC(F)(F)F)C(=O)N

Specifica

Materiale o nome chimico 2-Methyl-5-(trifluoromethoxy)benzamide
Quantità 1 g
Peso formulazione 219.16
Percent Purity 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'