Learn More
Thermo Scientific Chemicals 4mm Die Set for KBr Quick Press Kit
Codice prodotto. 15451557
Click to view available options
Dimensione campione:
2 mm
4 mm
7 mm
Dimensione della confezione:
Pezzo
Codice prodotto. 15451557
Marca: Thermo Scientific Chemicals 039788.KT
Richiesta di formato massivo o personalizzato
For use with KBr Quick Press Kit
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifica
Tipo Die Set
Dimensione campione 4 mm
Da utilizzare con (apparecchiatura) KBr Quick Press Kit