Learn More
3,5-dichlorobenzene-1-carbohydrazide, 97%, Thermo Scientific™
Product Code. 10585954
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10585954 1 g Amber Glass Bottle 1g
10026384 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10585954 Supplier Thermo Scientific Maybridge Supplier No. TL00894DA
 Restricted Item
Spedizione stimata: 30-11-2026
Add to basket
Chemical Identifiers
CAS 62899-78-9
Molecular Formula C7H6Cl2N2O
Molecular Weight (g/mol) 205.04
MDL Number MFCD00833411
InChI Key DNISXKBKTSHNKL-UHFFFAOYSA-N
Synonym 3,5-dichlorobenzhydrazide, 3,5-dichlorobenzenecarbohydrazide, acmc-1b4zh, 3,5-bis chloranyl benzohydrazide, 3,5-dichlorobenzoic acid hydrazide, benzoic acid,3,5-dichloro-,hydrazide, benzoic acid,3,5-dichloro-, hydrazide
PubChem CID 2735966
IUPAC Name 3,5-dichlorobenzohydrazide
SMILES NNC(=O)C1=CC(Cl)=CC(Cl)=C1
Specifications
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 205.04
Percent Purity 97%
Chemical Name or Material 3, 5-dichlorobenzene-1-carbohydrazide