Learn More
3-Fluorobenzotrifluoride, 99%
Product Code. 11497327
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11497327 25 g 25g
11407337 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11497327 Supplier Thermo Scientific Alfa Aesar Supplier No. A15055.14
 Restricted Item
In stock
Add to basket
CAS: 401-80-9 | C7H4F4 | 164.10 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 401-80-9
Molecular Formula C7H4F4
Molecular Weight (g/mol) 164.10
MDL Number MFCD00000382
InChI Key GBOWGKOVMBDPJF-UHFFFAOYSA-N
Synonym 3-fluorobenzotrifluoride, m-fluorobenzotrifluoride, 1-fluoro-3-trifluoromethyl benzene, alpha,alpha,alpha,3-tetrafluorotoluene, benzene, 1-fluoro-3-trifluoromethyl, trifluoromethyl-3-fluorobenzene, a,a,a,3-tetrafluorotoluene, 1-trifluoromethyl-3-fluorobenzene, 1-fluoro-3-trifluoro-methyl benzene, 3-trifluoromethyl fluorobenzene
PubChem CID 67867
IUPAC Name 1-fluoro-3-(trifluoromethyl)benzene
SMILES FC1=CC=CC(=C1)C(F)(F)F
Specifications
Density 1.303
Boiling Point 101°C to 102°C
Flash Point 7°C (44°F)
Refractive Index 1.401
Quantity 25 g
UN Number UN1993
Beilstein 2045664
Formula Weight 164.1
Percent Purity 99%
Chemical Name or Material 3-Fluorobenzotrifluoride