Learn More
Sampling Systems™ 316 Stainless Steel Storage Containers without Toggle Clamps
Multi-purpose containers with light duty construction for a wide range of storage uses in the laboratory.
Marca: Sampling Systems™ A466-25
Dettagli aggiuntivi : Peso : 18.14400kg
Descrizione
- Crevice-free interior
- Two sturdy handles
- Close fitting recessed lid with handle
- Flat bottom with chime ring to give added protection
- Toggle clamps and seals are optional extras
Specifica
Storage container | |
Recessed Lid | |
Stainless Steel | |
300 mm |
Yes | |
25 L | |
390 mm |