Learn More
Hamilton™ 33 gauge, Hubless Needle, custom length (0.375 to 12 in), point style 2, 3, or 4, 6/PK
Codice prodotto. 12311828
Click to view available options
Calibro dell'ago:
33 G
Dimensione della confezione:
pezzi 6
Codice prodotto. 12311828
Marca: Hamilton™ 2203301
This category of needles comes without a hub so they cannot be connected to a Hamilton syringe. The point style and length of the tubing can be specified at the time of order.
Specification Sheet
  • Gauge – 33 gauge
  • Point Style – 2, 3, or 4
  • Needle Length – Custom Needle Length
  • Autoclavable – Yes
  • Sales Quantity – 6 needles per pack
  • Hub – No Hub
Specifica
Calibro dell'ago 33 G
Product Type Hubless Needles