Learn More
Brannan™ ECO-therm Lab Thermometer
Codice prodotto. 15472685
Click to view available options
Temperature Range (Metric):
-10°C to +110°C
-10°C to +150°C
-35°C to +50°C
Tipo di immersione:
Partial Immersion
Total Immersion
Dimensione della confezione:
pezzi 10
Codice prodotto. 15472685
Marca: Brannan™ 46/706/0
Green spirit thermometers filled with non toxic expansion liquid.
  • Quality and resolution make these ideal substitute for general purpose mercury thermometers
  • Each thermometer is fitted with Brannan non roll end cap
  • Cap has integral ring to allow the thermometer to be suspended and is supplied in protective PVC packaging
Specifica
Tipo di immersione Partial Immersion
Lunghezza (sistema metrico) 305 mm
Materiale Glass
Riempimento termometro Green Spirit
Product Type Laboratory Immersion Thermometer
Temperatura max (sistema metrico) 150°C
Min. Temperature (Metric) -10°C
Temperature Range (Metric) -10°C to +150°C
Accuracy ±1.5°C
Suddivisione 1°C