Learn More
4-Bromo-3-chlorobenzotrifluoride, 98%
Product Code. 15498568
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15498568 5 g 5g
15408578 25 g 25g
15418578 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15498568 Supplier Thermo Scientific Alfa Aesar Supplier No. H61978.06
 Restricted Item
Spedizione stimata: 15-12-2026
Add to basket
CAS: 402-04-0 | C7H3BrClF3 | 259.45 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 402-04-0
Molecular Formula C7H3BrClF3
Molecular Weight (g/mol) 259.45
MDL Number MFCD00673985
InChI Key RVTIHGGJJMXISV-UHFFFAOYSA-N
Synonym 4-bromo-3-chlorobenzotrifluoride, 1-bromo-2-chloro-4-trifluoromethyl benzene, 1-bromo-2-chloro-4-trifluoromethyl-benzene, benzene, 1-bromo-2-chloro-4-trifluoromethyl, 3-chloro-4-bromobenzotrifluoride, 4-trifluoromethyl-2-chloro-bromo benzene, pubchem13478, ksc494s1t, 2-chloro-4-trifluoromethylbromobenzene, 4-cf3-2-chloro-bromo benzene
PubChem CID 14273266
IUPAC Name 1-bromo-2-chloro-4-(trifluoromethyl)benzene
SMILES C1=CC(=C(C=C1C(F)(F)F)Cl)Br
Specifications
Quantity 5 g
Formula Weight 259.45
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 4-Bromo-3-chlorobenzotrifluoride