Learn More
4-Methoxystilbene, 98%, Thermo Scientific Chemicals
Product Code. 11463567
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11463567 5 g 5g
11473567 25 g 25g
11483567 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11463567 Supplier Thermo Scientific Alfa Aesar Supplier No. A17328.06
 Restricted Item
In stock
Aggiungi al carrello
Aggiungi al carrello
Request Bulk or Custom Format
CAS: 1142-15-0 | Molecular Formula: C15H14O | Molecular Weight: 210.276 g/mol
It is used in the modification of the photochemical behavior of organic molecules by cyclodextrin.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used in the modification of the photochemical behavior of organic molecules by cyclodextrin.
Solubility
Sparingly soluble (0.061 g/L) (25°C) in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY
Identificatori chimici
CAS 1142-15-0
Molecular Formula C15H14O
Molecular Weight (g/mol) 210.276
MDL Number MFCD00017178
InChI Key XWYXLYCDZKRCAD-BQYQJAHWSA-N
Synonym p-methoxystilbene, 4-methoxystilbene, anisole, p-styryl, trans-4-methoxystilbene, e-4-methoxystilbene, 1-methoxy-4-e-2-phenylethenyl benzene, 1-p-methoxyphenyl-2-phenylethene, unii-5el148b7hg, benzene, 1-methoxy-4-2-phenylethenyl, 1-methoxy-4-e-styryl-benzene
PubChem CID 1245920
ChEBI CHEBI:35094
IUPAC Name 1-methoxy-4-[(E)-2-phenylethenyl]benzene
SMILES COC1=CC=C(C=C1)C=CC2=CC=CC=C2
Specifica
Melting Point 136°C to 138°C
Quantity 5 g
Solubility Information Sparingly soluble (0.061g/L) (25°C) in water.
Formula Weight 210.27
Percent Purity 98%
Chemical Name or Material 4-Methoxystilbene