Learn More
SHOWA™ 477 Series Nitrile Coated Cold Resistant Gloves
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
Dimensione della confezione:
5 paia
Descrizione
Benifits
- Seamless knit
- Cold Resistant
- Water-resistant
- Flexible
- Abrasion-resistant
- Extra Grip
- Thermal insulation
- Form-fitting
- Impermeable
Features
- Foam grip
- Elasticated wrist cuff
- Ergonomic
- Foam nitrile palm dip
- Insulated
- Fully-coated nitrile
Trades & Applications
- Concreting
- Guttering and drain pipe
- Insulation
- Washing and cleaning
- Planting & repotting
- Bottling
- Water/Sewage treatment
- Plumbing
- Drainage, piping
- Shipping/Receiving
Specifica
Specifica
| Certificazione/Conformità | CE Cat II, EN 388:2016, EN 511:2006 |
| Struttura | Seamless Knit |
| Coating Material | Nitrile |
| Colore | Blue, Black |
| Materiale | Nylon, Polyester |
| Size | 2X-Large |
| Materiale rivestimento interno | Acrylic |
| Resistente a | Cold Resistant, Water Resistant, Abrasion Resistant |
| Serie | 477 |