Learn More
Hamilton™ 1000 Series Gastight™ Saltline Syringes
Click to view available options
Volume (sistema metrico):
1 mL
5 mL
10 mL
Dimensione della confezione:
Pezzo
Descrizione
- Equipped with inert and non-corrosive plunger
- Fitted with resistant ultra-high molecular weight poly-ethylene (UHMWPE) plunger tip
Applications:
For life science applications with salty buffers, mining applications with acids or trace metal analysis
Specifica
Specifica
| Autoclavabile | Not Autoclavable |
| Product Type | Gastight Saltline Syringe |
| Volume (sistema metrico) | 10 mL |
| Tipo di collegamento | Bubble Free Prime (BFP) |