Learn More
Hamilton™ Microlab™ 1700 Series Gastight™ Syringes
Click to view available options
Volume (sistema metrico):
10 μL
25 μL
100 μL
Calibro dell'ago:
22s G
26s G
Modello:
1701
1702
1710
Dimensione della confezione:
Pezzo
Descrizione
- Equipped with precision-machined polymer plunger tip, often PTFE, which creates leak-free seal
- Tight fit feature ensures tip wipes interior of syringe barrel free of sample
- Reduces the chance deposit will occur in samples with dissolved or suspended molecules and result in damaged plunger
- ChemSeal termination
Specifica
Specifica
| Calibro dell'ago | 22s G |
| Modello | 1710 |
| Volume (sistema metrico) | 100 μL |