Learn More
Hamilton™ Microlab™ 1700 Series Gastight™ Syringes
Codice prodotto. 11542385
Click to view available options
Volume (sistema metrico):
10 μL
25 μL
100 μL
Calibro dell'ago:
22s G
26s G
Modello:
1701
1702
1710
Dimensione della confezione:
Pezzo
Codice prodotto. 11542385
Marca: Hamilton™ 80262
For dispensing both liquids and gases
  • Equipped with precision-machined polymer plunger tip, often PTFE, which creates leak-free seal
  • Tight fit feature ensures tip wipes interior of syringe barrel free of sample
  • Reduces the chance deposit will occur in samples with dissolved or suspended molecules and result in damaged plunger
  • ChemSeal termination
Specifica
Calibro dell'ago 22s G
Modello 1702
Volume (sistema metrico) 25 μL