Learn More
Invitrogen™ 4EBP1 Monoclonal Antibody (4B6G10)
Mouse Monoclonal Antibody
Marca: Invitrogen™ MA515313
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
MA5-15313 targets 4E-BP1 in indirect ELISA, IHC and WB applications and shows reactivity with Human samples. The MA5-15313 immunogen is purified recombinant fragment of 4E-BP1 expressed in E. Coli.
This gene encodes one member of a family of translation repressor proteins. The protein directly interacts with eukaryotic translation initiation factor 4E, which is a limiting component of the multisubunit complex that recruits 40S ribosomal subunits to the 5' end of mRNAs. Interaction of this protein with eIF4E inhibits complex assembly and represses translation. This protein is phosphorylated in response to various signals including UV irradiation and insulin signaling, resulting in its dissociation from eIF4E and activation of mRNA translation.
Specifica
| 4EBP1 | |
| Monoclonal | |
| Conc. Not Determined | |
| ascites with 0.03% sodium azide | |
| Q13541 | |
| EIF4EBP1 | |
| Purified recombinant fragment of 4E-BP1 expressed in E. Coli. | |
| RUO | |
| 1978 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 4B6G10 | |
| Unconjugated | |
| EIF4EBP1 | |
| 4EBP1; 4E-BP1; AA959816; BP-1; eIF4E-binding protein 1; EIF4EBP1; eukaryotic translation initiation factor 4E binding protein 1; eukaryotic translation initiation factor 4E-binding protein 1; MGC431; MGC4316; P/OKCL.6; PHAS-1; PHAS-I; Phosphorylated heat- and acid-stable protein regulated by insulin 1 | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |