Learn More
Agilent BioTek 50 TS Microplate Washer
GREENER_CHOICE
Codice prodotto. 17237789 Sfoglia Tutto Agilent BioTek Prodotti
Click to view available options
Modello:
50TS12
50TS12V
50TS16
50TS16V
50TS8
50TS8F
50TS8M
50TS8MF
50TS8MV
50TS8V
Dimensione della confezione:
Pezzo
Codice prodotto. 17237789
Marca: Agilent BioTek 50TS16VSN
Questo articolo richiede un aggiornamento del prezzo prima di essere ordinato.
Richiedi qui il prezzo per questo articolo.
The BioTek 50 TS washer offers excellent microplate washing performance. For Research Use Only. Not for use in diagnostic procedures.
The BioTek 50 TS washer offers excellent microplate washing performance. Ideal for use as an ELISA washer, the system is also well suited for cell-based assay washing. The 50 TS can process biomagnetic or polystyrene bead and vacuum filtration protocols as well
The color touch screen provides a visual interface that makes creating protocols fast and intuitive. When used with the 800 TS absorbance reader or other detection system, the 50 TS delivers convenience and high-quality results to your laboratory plate washing workflows.
Warranty
12 months
TRUSTED_SUSTAINABILITY
Specifica
Certificazione/Conformità CE and TUV marked. RoHS compliant.
Collegamenti USB (1)
Bottiglie Optional (2 to 4 L)
Tipo di piastra 24-, 96-, 384-well Plates (96- and 384-well: Standard and Low Profile Plates, Solid and Filter Bottom (Option), 24-well Plates with 4-well Manifold)
Tempo di immersione Programmable in minutes and seconds, up to 30 min.
Tipo Microplate Washer
Peso (sistema metrico) 9.8 kg
Cicli di lavaggio 1 to 10
Volume di lavaggio 25 to 3000 μL/well
Display 4.3 in. LCD Color Touch Screen
Portata 150 to 1000 μL/well/sec. (manifold dependent)
Tipo di erogazione del fluido One positive displacement syringe drive
Modello 50TS16V
Include 96-/384-well plates; Buffer Switching
Interfaccia di automazione 4.3 in. Color LCD Touch Screen Display
Peso (sistema imperiale) 22 lb.