missing translation for 'onlineSavingsMsg'
Learn More

Phenylephrine hydrochloride, MedChemExpress

Codice prodotto. 16482146
Encompass_Preferred
Click to view available options
Quantità:
100 mg
500 mg
Dimensione della confezione:
100mg
500mg
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Codice prodotto. 16482146

Marca: Medchem Express HYB0471

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

MedChemExpress (R)-(-)-Phenylephrine hydrochloride is a selective α1-adrenoceptor agonist with pKis of 5.86, 4.87 and 4.70 for α1D, α1B and α1A receptors respectively.

(R)-(-)-Phenylephrine hydrochloride is a selective α1-adrenoceptor agonist with pKis of 5.86, 4.87 and 4.70 for α1D, α1B and α1A receptors respectively.

Specifica

Materiale o nome chimico Phenylephrine hydrochloride
CAS 61-76-7
Colore White
Note sul grado di purezza Research
Formula molecolare C9H14ClNO2
Quantità 500 mg
Sinonimo (R)-(-)-Phenylephrine hydrochlorideL-Phenylephrine hydrochloride
Informazioni di solubilità DMSO : ≥ 150 mg/mL (736.49 mM) ∣H2O : 100 mg/mL (490.99 mM; Need ultrasonic)
SMILES OC1=CC([C@@H](O)CNC)=CC=C1.Cl
Molecular Weight (g/mol) 203.67
Peso formulazione 203.67
Percent Purity 99.76%
Grado Research
Forma fisica Solid
Da utilizzare con (applicazione) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.