Learn More
Spectrum™ Labs 0.5 - 1.0 kD Biotech CE Dialysis Kits
Codice prodotto. p-5022198
Click to view available options
Diametro (sistema metrico):
6.4 mm
10 mm
15 mm
20 mm
Lunghezza (sistema metrico):
0.32 mL/cm vol./length
0.79 mL/cm vol./length
1.8 mL/cm vol./length
3.1 mL/cm vol./length
Dimensione della confezione:
1m
Pezzo
Codice prodotto. 15370662
Marca: Spectrum™ 131096T
Includes
  • 1m Biotech CE Tubing in 0.05% sodium azide
  • Two universal tubing closures
  • Five opening picks
With tubing made from biologically inert and ultra-pure Biotech cellulose ester membrane
Ideal for small, critical dialysis applications that require the more concise selectivity and retention provided by Biotech Cellulose Ester (CE) Membrane than Standard Grade RC
  • Hydrophilic, symmetric porositiy
  • For small tubing demand applications or evaluating a specific dialysis membrane
  • Includes 1m tubing with pair of tubing closures and 5 opening picks
Specifica
MWCO 0.5 to 1.0 kDa
Diametro (sistema metrico) 20 mm
Lunghezza (sistema metrico) 3.1 mL/cm vol./length
Quantità 1 m Tubing/Kit