Learn More
Spectrum™ Labs Spectra/Por™ 1 6-8 kD MWCO Standard RC Dry Dialysis Kits
Codice prodotto. 15350742
Click to view available options
Diametro (sistema metrico):
6.4 mm
14.6 mm
20.4 mm
25.5 mm
32 mm
64 mm
76 mm
Lunghezza (sistema metrico):
0.32 mL/cm vol./length
1.7 mL/cm
3.3 mL/cm vol./length
32 mL/cm vol./length
46 mL/cm vol./length
5 m
5.1 mL/cm vol./length
7.9 mL/cm vol./length
Quantità:
1 m Tubing/Kit
5 m Tubing/Kit
Each
Dimensione della confezione:
1m
5m
Codice prodotto. 15350742
Marca: Spectrum™ 132645T
Includes
  • 1m or 5m of Spectra/Por 1 dialysis tubing
  • 1 Standard Spectra/Por Closure (for buoyance)
  • 1 weighted Spectra/Por Closure (for orientation)
  • 2 adhesive tubing openers
With Standard Grade Regenerated Cellulose membrane tubing for applications with significant size difference between MW species being separated
  • For basic dialysis applications, or evaluating dialysis tubing for dialysis application
  • Packaged dry with glycerin (soak in water)
Specifica
MWCO 6 to 8 kDa
Diametro (sistema metrico) 6.4 mm
Lunghezza (sistema metrico) 0.32 mL/cm vol./length
Quantità 5 m Tubing/Kit